Assay Details
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007701.1 | NP_001007702.1 | 585 | 85 | ||
GenBank mRNA | AY387055.1 | - | 468 | 85 | ||
BC083851.1 | - | 585 | 85 | |||
FQ223598.1 | - | 457 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
