Assay Details
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007701.1 | NP_001007702.1 | 637 | 94 | ||
GenBank mRNA | AY387055.1 | - | 520 | 94 | ||
BC083851.1 | - | 637 | 94 | |||
FQ223598.1 | - | 509 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
