Assay Details
Gene Symbol: Ewsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025632.2 | NP_001020803.2 | 10 - 11 | 1075 | 87 | |
XM_006251201.2 | XP_006251263.2 | 10 - 11 | 1060 | 87 | ||
XM_006251202.2 | XP_006251264.2 | 10 - 11 | 1057 | 87 | ||
XM_006251203.2 | XP_006251265.2 | 10 - 11 | 1086 | 87 | ||
GenBank mRNA | BC162004.1 | - | 10 - 11 | 1075 | 87 |
Gene Symbol: LOC100912481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749683.2 | XP_003749731.1 | 10 - 11 | 1080 | 87 | |
XM_003749684.2 | XP_003749732.1 | 11 - 12 | 1095 | 87 | ||
XM_006236016.1 | XP_006236078.1 | 11 - 12 | 1098 | 87 | ||
XM_006236017.1 | XP_006236079.1 | 10 - 11 | 1077 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
