Assay Details
Gene Symbol: Ewsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025632.2 | NP_001020803.2 | 5 - 6 | 454 | 103 | |
XM_006251201.2 | XP_006251263.2 | 5 - 6 | 436 | 103 | ||
XM_006251202.2 | XP_006251264.2 | 5 - 6 | 436 | 103 | ||
XM_006251203.2 | XP_006251265.2 | 5 - 6 | 462 | 103 | ||
XM_008770260.1 | XP_008768482.1 | 5 - 6 | 462 | 103 | ||
GenBank mRNA | BC162004.1 | - | 5 - 6 | 454 | 103 |
Gene Symbol: LOC100912481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749683.2 | XP_003749731.1 | 5 - 6 | 456 | 103 | |
XM_003749684.2 | XP_003749732.1 | 6 - 7 | 474 | 103 | ||
XM_006236016.1 | XP_006236078.1 | 6 - 7 | 474 | 103 | ||
XM_006236017.1 | XP_006236079.1 | 5 - 6 | 456 | 103 | ||
XM_008762731.1 | XP_008760953.1 | 5 - 6 | 456 | 103 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
