Assay Details
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019328.3 | NP_062201.2 | 5 - 6 | 1474 | 106 | |
XM_006234201.2 | XP_006234263.1 | 6 - 7 | 1692 | 106 | ||
XM_008761861.1 | XP_008760083.1 | 5 - 6 | 1608 | 106 | ||
GenBank mRNA | DQ092633.1 | - | 5 - 6 | 1241 | 106 | |
U01146.1 | - | 5 - 6 | 1405 | 106 | ||
U72345.1 | - | 6 - 7 | 1698 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
