Assay Details
Gene Symbol: Psmb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053532.1 | NP_445984.1 | 6 - 7 | 582 | 99 | |
GenBank mRNA | AF285103.1 | - | 6 - 7 | 582 | 99 | |
BC060551.1 | - | 6 - 7 | 592 | 99 | ||
FQ210403.1 | - | 5 - 6 | 475 | 99 | ||
FQ212825.1 | - | 5 - 6 | 475 | 99 | ||
FQ213190.1 | - | 5 - 6 | 475 | 99 | ||
FQ215265.1 | - | 5 - 6 | 475 | 99 | ||
FQ216419.1 | - | 5 - 6 | 475 | 99 | ||
FQ218632.1 | - | 5 - 6 | 475 | 99 | ||
FQ219494.1 | - | 5 - 6 | 475 | 99 | ||
FQ219779.1 | - | 5 - 6 | 475 | 99 | ||
FQ220023.1 | - | 5 - 6 | 475 | 99 | ||
FQ229165.1 | - | 5 - 6 | 475 | 99 | ||
FQ229238.1 | - | 5 - 6 | 475 | 99 | ||
FQ229410.1 | - | 5 - 6 | 475 | 99 | ||
FQ229570.1 | - | 5 - 6 | 475 | 99 | ||
FQ229735.1 | - | 5 - 6 | 475 | 99 | ||
FQ229750.1 | - | 5 - 6 | 475 | 99 | ||
FQ232691.1 | - | 5 - 6 | 475 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
