Assay Details
Gene Symbol: Mmadhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004280.1 | NP_001004280.1 | 6 - 7 | 716 | 84 | |
XM_006234158.2 | XP_006234220.1 | 6 - 7 | 835 | 84 | ||
GenBank mRNA | BC078953.1 | - | 6 - 7 | 716 | 84 | |
FQ212704.1 | - | 6 - 7 | 711 | 84 | ||
FQ216239.1 | - | 6 - 7 | 711 | 84 | ||
FQ219117.1 | - | 6 - 7 | 712 | 84 | ||
FQ228704.1 | - | 6 - 7 | 712 | 84 | ||
FQ229971.1 | - | 6 - 7 | 681 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
