Assay Details
Gene Symbol: Mmadhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004280.1 | NP_001004280.1 | 1 - 2 | 55 | 73 | |
GenBank mRNA | BC078953.1 | - | 1 - 2 | 55 | 73 | |
FQ212704.1 | - | 1 - 2 | 50 | 73 | ||
FQ216239.1 | - | 1 - 2 | 50 | 73 | ||
FQ219117.1 | - | 1 - 2 | 51 | 73 | ||
FQ228704.1 | - | 1 - 2 | 51 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC078953.1, NM_001004280.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
