Assay Details
Gene Symbol: Epc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108581.1 | NP_001102051.1 | 4 - 5 | 741 | 86 | |
XM_006234155.2 | XP_006234217.1 | 4 - 5 | 715 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
