Assay Details
Gene Symbol: Acvr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024486.1 | NP_077812.1 | 2 - 3 | 216 | 80 | |
XM_006234214.2 | XP_006234276.1 | 4 - 5 | 1821 | 80 | ||
GenBank mRNA | BC167754.1 | - | 3 - 4 | 387 | 80 | |
L19341.1 | - | 2 - 3 | 216 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
