Assay Details
Gene Symbol: Spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012881.2 | NP_037013.2 | 2 - 3 | 161 | 95 | |
XM_008769996.1 | XP_008768218.1 | 2 - 3 | 234 | 95 | ||
GenBank mRNA | AB001382.1 | - | 1 - 2 | 55 | 95 | |
BC078874.1 | - | 2 - 3 | 141 | 95 | ||
CV102664.1 | - | 2 - 3 | 161 | 95 | ||
FQ213473.1 | - | 2 - 3 | 150 | 95 | ||
FQ222192.1 | - | 2 - 3 | 177 | 95 | ||
M99252.1 | - | 2 - 3 | 170 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
