Assay Details
Gene Symbol: Mrps18c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105996.1 | NP_001099466.1 | 4 - 5 | 328 | 135 | |
GenBank mRNA | FQ224447.1 | - | 461 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
