Assay Details
Gene Symbol: Enoph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009391.1 | NP_001009391.1 | 3 - 4 | 479 | 63 | |
XM_008770023.1 | XP_008768245.1 | 3 - 4 | 529 | 63 | ||
GenBank mRNA | BC087697.1 | - | 3 - 4 | 479 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
