Assay Details
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030849.1 | NP_110476.1 | 2 - 3 | 295 | 81 | |
GenBank mRNA | D17667.1 | - | 1 - 2 | 70 | 81 | |
D38082.1 | - | 1 - 2 | 152 | 81 | ||
S75359.1 | - | 2 - 3 | 295 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
