Assay Details
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030849.1 | NP_110476.1 | 4 - 5 | 559 | 65 | |
GenBank mRNA | D17667.1 | - | 3 - 4 | 334 | 65 | |
D38082.1 | - | 3 - 4 | 416 | 65 | ||
S75359.1 | - | 4 - 5 | 559 | 65 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
