Assay Details
Gene Symbol: Arhgap22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107297.1 | NP_001100767.1 | 2 - 3 | 294 | 80 | |
XM_006252749.2 | XP_006252811.1 | 2 - 3 | 224 | 80 | ||
XM_006252750.2 | XP_006252812.1 | 2 - 3 | 804 | 80 | ||
XM_008771098.1 | XP_008769320.1 | 4 - 5 | 950 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P068548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86509 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P068548 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P068548 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P068548 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1041539 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10786862 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10786863 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10786861 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10786855 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10786856 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10786858 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10786864 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10786851 | - | - |
Rat Genome 230 2.0 Array | 1391083_at | - | - |
Rat Genome 230 2.0 Array | 1391083_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391083_at | - | - |
Affymetrix HT Rat Focus Array | 1391083_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600001467.rn.1 | TC1600000127.rn.1 | RTA TC |
Rat Expression Set 230 | 1391083_at | - | - |
Rat Expression Set 230 | 1391083_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10786851 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21023724 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353116 | - | - |
