Assay Details
Gene Symbol: Glt8d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007683.1 | NP_001007684.1 | 6 - 7 | 712 | 94 | |
GenBank mRNA | AY325256.1 | - | 6 - 7 | 659 | 94 | |
BC079066.1 | - | 6 - 7 | 712 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
