Assay Details
Gene Symbol: Il17rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107290.1 | NP_001100760.1 | 6 - 7 | 628 | 110 | |
XM_006252603.2 | XP_006252665.1 | 6 - 7 | 610 | 110 | ||
GenBank mRNA | BC168886.1 | - | 2 - 3 | 173 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
