Assay Details
Gene Symbol: Mettl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007623.1 | NP_001007624.1 | 2 - 3 | 465 | 70 | |
XM_006252582.2 | XP_006252644.1 | 1 - 2 | 360 | 70 | ||
XM_006252583.2 | XP_006252645.1 | 1 - 2 | 363 | 70 | ||
XM_008770988.1 | XP_008769210.1 | 1 - 2 | 360 | 70 | ||
GenBank mRNA | BC079309.1 | - | 2 - 3 | 465 | 70 | |
FQ211422.1 | - | 2 - 3 | 473 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
