Assay Details
Gene Symbol: Sirt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004256.1 | NP_001004256.1 | 1 - 2 | 163 | 94 | |
XM_006253801.1 | XP_006253863.1 | 2 - 3 | 347 | 94 | ||
XM_006253802.1 | XP_006253864.1 | 2 - 3 | 225 | 94 | ||
XM_006253803.2 | XP_006253865.1 | 2 - 3 | 219 | 94 | ||
XM_006253804.2 | XP_006253866.1 | 2 - 3 | 170 | 94 | ||
GenBank mRNA | BC078958.1 | - | 1 - 2 | 163 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
