Assay Details
Gene Symbol: Gmds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039606.1 | NP_001034695.1 | 3 - 4 | 261 | 127 | |
GenBank mRNA | BC104708.1 | - | 3 - 4 | 261 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
