Assay Details
Gene Symbol: Ndufs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230731.1 | XP_006230793.1 | 2 - 3 | 302 | 144 | |
GenBank mRNA | FQ213239.1 | - | 2 - 3 | 302 | 144 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006230731.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
