Assay Details
Gene Symbol: Spata6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014165.1 | NP_001014187.1 | 6 - 7 | 595 | 86 | |
XM_006231237.2 | XP_006231299.1 | 6 - 7 | 554 | 86 | ||
XM_006231239.2 | XP_006231301.1 | 6 - 7 | 471 | 86 | ||
XM_006231240.2 | XP_006231302.1 | 5 - 6 | 441 | 86 | ||
XM_008760342.1 | XP_008758564.1 | 7 - 8 | 624 | 86 | ||
XM_008760343.1 | XP_008758565.1 | 7 - 8 | 625 | 86 | ||
GenBank mRNA | BC079024.1 | - | 6 - 7 | 595 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
