Assay Details
Gene Symbol: Spata6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014165.1 | NP_001014187.1 | 10 - 11 | 1146 | 98 | |
XM_006231237.2 | XP_006231299.1 | 10 - 11 | 1105 | 98 | ||
XM_006231239.2 | XP_006231301.1 | 10 - 11 | 1022 | 98 | ||
XM_006231240.2 | XP_006231302.1 | 9 - 10 | 992 | 98 | ||
XM_008760342.1 | XP_008758564.1 | 11 - 12 | 1175 | 98 | ||
XM_008760343.1 | XP_008758565.1 | 11 - 12 | 1176 | 98 | ||
GenBank mRNA | BC079024.1 | - | 10 - 11 | 1146 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
