Assay Details
Gene Symbol: Polr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053948.4 | NP_446400.1 | 4 - 5 | 404 | 112 | |
XM_006230973.1 | XP_006231035.1 | 6 - 7 | 689 | 112 | ||
GenBank mRNA | BC060550.1 | - | 4 - 5 | 360 | 112 | |
BC060595.1 | - | 6 - 7 | 698 | 112 | ||
FQ223906.1 | - | 6 - 7 | 694 | 112 | ||
FQ224086.1 | - | 4 - 5 | 437 | 112 | ||
FQ230390.1 | - | 4 - 5 | 367 | 112 | ||
FQ232354.1 | - | 4 - 5 | 408 | 112 | ||
Z71925.1 | - | 4 - 5 | 405 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
