Assay Details
Gene Symbol: Lipa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012732.3 | NP_036864.3 | 1 - 2 | 145 | 58 | |
GenBank mRNA | BC072532.1 | - | 1 - 2 | 145 | 58 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC072532.1, NM_012732.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
