Assay Details
Gene Symbol: Oat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022521.3 | NP_071966.1 | 1 - 2 | 70 | 70 | |
GenBank mRNA | BC061551.1 | - | 1 - 2 | 73 | 70 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC061551.1, NM_022521.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
