Assay Details
Gene Symbol: Cd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019295.1 | NP_062168.1 | 4 - 5 | 549 | 79 | |
GenBank mRNA | BC098918.1 | - | 4 - 5 | 556 | 79 | |
D10728.1 | - | 4 - 5 | 549 | 79 | ||
X78985.1 | - | 4 - 5 | 496 | 79 |
Gene Symbol: LOC100911215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231081.2 | XP_006231143.2 | 370 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
