Assay Details
Gene Symbol: Spata6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014165.1 | NP_001014187.1 | 1 - 2 | 161 | 60 | |
XM_008760342.1 | XP_008758564.1 | 2 - 3 | 190 | 60 | ||
XM_008760343.1 | XP_008758565.1 | 2 - 3 | 191 | 60 | ||
GenBank mRNA | BC079024.1 | - | 1 - 2 | 161 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
