Assay Details
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014254.1 | NP_001014276.1 | 11 - 12 | 1495 | 81 | |
XM_008761185.1 | XP_008759407.1 | 11 - 12 | 1700 | 81 | ||
XM_008761186.1 | XP_008759408.1 | 12 - 13 | 1567 | 81 | ||
XM_008761187.1 | XP_008759409.1 | 11 - 12 | 1697 | 81 | ||
XM_008761188.1 | XP_008759410.1 | 11 - 12 | 1679 | 81 | ||
XM_008761189.1 | XP_008759411.1 | 11 - 12 | 1676 | 81 | ||
XM_008761190.1 | XP_008759412.1 | 11 - 12 | 1676 | 81 | ||
XM_008761191.1 | XP_008759413.1 | 11 - 12 | 1673 | 81 | ||
XM_008761192.1 | XP_008759414.1 | 11 - 12 | 1546 | 81 | ||
XM_008761193.1 | XP_008759415.1 | 8 - 9 | 1347 | 81 | ||
XM_008761194.1 | XP_008759416.1 | 8 - 9 | 826 | 81 | ||
GenBank mRNA | BC083788.1 | - | 11 - 12 | 1495 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
