Assay Details
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014254.1 | NP_001014276.1 | 1 - 2 | 264 | 90 | |
XM_008761192.1 | XP_008759414.1 | 1 - 2 | 294 | 90 | ||
GenBank mRNA | BC083788.1 | - | 1 - 2 | 264 | 90 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC083788.1, NM_001014254.1, XM_008761192.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
