Assay Details
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014254.1 | NP_001014276.1 | 4 - 5 | 755 | 83 | |
XM_008761185.1 | XP_008759407.1 | 4 - 5 | 960 | 83 | ||
XM_008761186.1 | XP_008759408.1 | 5 - 6 | 827 | 83 | ||
XM_008761187.1 | XP_008759409.1 | 4 - 5 | 957 | 83 | ||
XM_008761188.1 | XP_008759410.1 | 4 - 5 | 939 | 83 | ||
XM_008761189.1 | XP_008759411.1 | 4 - 5 | 936 | 83 | ||
XM_008761190.1 | XP_008759412.1 | 4 - 5 | 936 | 83 | ||
XM_008761191.1 | XP_008759413.1 | 4 - 5 | 933 | 83 | ||
XM_008761192.1 | XP_008759414.1 | 4 - 5 | 806 | 83 | ||
GenBank mRNA | BC083788.1 | - | 4 - 5 | 755 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
