Assay Details
Gene Symbol: Mrpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106903.1 | NP_001100373.1 | 5 - 6 | 440 | 65 | |
XM_008766999.1 | XP_008765221.1 | 4 - 5 | 399 | 65 | ||
GenBank mRNA | BC168851.1 | - | 4 - 5 | 335 | 65 | |
FQ224516.1 | - | 4 - 5 | 332 | 65 |
Gene Symbol: NEWGENE_1308196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767056.1 | XP_008765278.1 | 437 | 65 | ||
XM_008767057.1 | XP_008765279.1 | 519 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
