Assay Details
Gene Symbol: Trpv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053686.1 | NP_446138.1 | 7 - 8 | 1198 | 116 | |
XM_006236375.2 | XP_006236437.1 | 7 - 8 | 814 | 116 | ||
GenBank mRNA | AF160798.1 | - | 7 - 8 | 1198 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
