Assay Details
Gene Symbol: Eef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017245.2 | NP_058941.1 | 5 - 6 | 852 | 135 | |
GenBank mRNA | BC066661.1 | - | 5 - 6 | 852 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
