Assay Details
Gene Symbol: Csnk1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033870.1 | NP_001029042.1 | 10 - 11 | 1544 | 75 | |
NM_023102.2 | NP_075590.2 | 10 - 11 | 1541 | 75 | ||
XM_006241000.2 | XP_006241062.1 | 9 - 10 | 2919 | 75 | ||
XM_006241001.2 | XP_006241063.1 | 9 - 10 | 1627 | 75 | ||
XM_006241002.2 | XP_006241064.1 | 10 - 11 | 2839 | 75 | ||
XM_006241003.2 | XP_006241065.1 | 9 - 10 | 2269 | 75 | ||
XM_008765130.1 | XP_008763352.1 | 9 - 10 | 1627 | 75 | ||
GenBank mRNA | BC072533.1 | - | 10 - 11 | 1544 | 75 | |
BC099758.1 | - | 4 - 5 | 403 | 75 | ||
U22297.1 | - | 9 - 10 | 1275 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
