Assay Details
Gene Symbol: Ndufs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008525.1 | NP_001008525.1 | 4 - 5 | 251 | 69 | |
XM_006240972.2 | XP_006241034.1 | 5 - 6 | 498 | 69 | ||
GenBank mRNA | BC086574.1 | - | 4 - 5 | 251 | 69 | |
FQ232492.1 | - | 4 - 5 | 260 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
