Assay Details
Gene Symbol: Pigl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138901.2 | NP_620256.1 | 5 - 6 | 539 | 60 | |
XM_008767767.1 | XP_008765989.1 | 5 - 6 | 535 | 60 | ||
GenBank mRNA | BC074020.1 | - | 5 - 6 | 539 | 60 | |
D88364.1 | - | 5 - 6 | 920 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
