Assay Details
Gene Symbol: Tmem98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007672.1 | NP_001007673.1 | 4 - 5 | 428 | 76 | |
GenBank mRNA | BC078932.1 | - | 4 - 5 | 428 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
