Assay Details
Gene Symbol: Mlx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034112.1 | NP_001029284.1 | 5 - 6 | 535 | 122 | |
XM_006247362.2 | XP_006247424.1 | 5 - 6 | 433 | 122 | ||
GenBank mRNA | BC128702.1 | - | 5 - 6 | 569 | 122 | |
CO560831.1 | - | 5 - 6 | 535 | 122 | ||
DQ350891.1 | - | 5 - 6 | 372 | 122 | ||
DQ350892.1 | - | 5 - 6 | 372 | 122 | ||
DQ350893.1 | - | 5 - 6 | 372 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
