Assay Details
Gene Symbol: Mrpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_573060.5 | XP_573060.3 | 4 - 5 | 733 | 74 | |
GenBank mRNA | FQ219872.1 | - | 4 - 5 | 705 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
