Assay Details
Gene Symbol: Skap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173311.2 | NP_775433.2 | 1 - 2 | 147 | 108 | |
GenBank mRNA | BC097931.1 | - | 1 - 2 | 147 | 108 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC097931.1, NM_173311.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
