Assay Details
Gene Symbol: Skap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173311.2 | NP_775433.2 | 4 - 5 | 378 | 80 | |
GenBank mRNA | AB092812.1 | - | 4 - 5 | 280 | 80 | |
BC097931.1 | - | 4 - 5 | 378 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
