Assay Details
Gene Symbol: Stk32b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107224.2 | NP_001100694.1 | 1102 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P031335 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P031329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55696 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031335 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031329 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031329 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031335 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P031329 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P031335 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P208427 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83122 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79370 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49862 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90545 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98888 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52144 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10773265 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773258 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773256 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773260 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773261 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773262 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773264 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773257 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773263 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10773255 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10773259 | - | - |
Affymetrix Rat Clariom S Assay | PSR1400005330.rn.1 | TC1400000777.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400005329.rn.1 | TC1400000777.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10773259 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773255 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371111 | - | - |
