Assay Details
Gene Symbol: Pisd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251423.2 | XP_006251485.1 | 3 - 4 | 4281 | 111 | |
XM_006251424.2 | XP_006251486.1 | 3 - 4 | 3463 | 111 | ||
XM_006251426.2 | XP_006251488.1 | 2 - 3 | 4045 | 111 | ||
XM_008770389.1 | XP_008768611.1 | 2 - 3 | 3590 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
