Assay Details
Gene Symbol: Sh3bp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054011.1 | NP_446463.2 | 1 - 2 | 146 | 81 | |
GenBank mRNA | AB027562.1 | - | 1 - 2 | 146 | 81 | |
FQ210473.1 | - | 1 - 2 | 273 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
