Assay Details
Gene Symbol: Sh3bp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054011.1 | NP_446463.2 | 6 - 7 | 674 | 102 | |
XM_008770976.1 | XP_008769198.1 | 4 - 5 | 2536 | 102 | ||
GenBank mRNA | AB027562.1 | - | 6 - 7 | 674 | 102 | |
FQ210473.1 | - | 7 - 8 | 894 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
