Assay Details
Gene Symbol: Oxnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107295.1 | NP_001100765.1 | 2 - 3 | 315 | 71 | |
XM_006252623.2 | XP_006252685.1 | 2 - 3 | 416 | 71 | ||
XM_006252624.2 | XP_006252686.1 | 3 - 4 | 523 | 71 | ||
XM_006252625.2 | XP_006252687.1 | 3 - 4 | 509 | 71 | ||
XM_006252626.2 | XP_006252688.1 | 2 - 3 | 402 | 71 | ||
XM_008771000.1 | XP_008769222.1 | 2 - 3 | 416 | 71 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008771000.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
