Assay Details
Gene Symbol: Oxnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107295.1 | NP_001100765.1 | 3 - 4 | 384 | 106 | |
XM_006252623.2 | XP_006252685.1 | 3 - 4 | 485 | 106 | ||
XM_006252624.2 | XP_006252686.1 | 4 - 5 | 592 | 106 | ||
XM_006252625.2 | XP_006252687.1 | 4 - 5 | 578 | 106 | ||
XM_006252626.2 | XP_006252688.1 | 3 - 4 | 471 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
