Assay Details
Gene Symbol: Slc7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013111.3 | NP_037243.2 | 1447 | 96 | ||
XM_008768943.1 | XP_008767165.1 | 6 - 7 | 1719 | 96 | ||
XM_008768944.1 | XP_008767166.1 | 6 - 7 | 1719 | 96 | ||
GenBank mRNA | AB066224.1 | - | 6 - 7 | 1204 | 96 | |
AF245000.1 | - | 1387 | 96 | |||
D67087.1 | - | 6 - 7 | 1177 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
